Anti-CD9 Mouse Monoclonal Antibody [clone: 8J244]

Lieferant: United States Biological
USBIC2260-09DEA 409 EUR
USBIC2260-09D
Anti-CD9 Mouse Monoclonal Antibody [clone: 8J244]
Antikörper
Recommended dilution: Western Blot: 2-4?g/ml

Type: Primary
Antigen: CD9 (CD9 molecule)
Clonality: Monoclonal
Clone: 8J244
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR