Anti-CD8A Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 3H885]

Lieferant: United States Biological
USBIC2259-97E3EA 0 EUR
USBIC2259-97E3
Anti-CD8A Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 3H885]
Antikörper
Recommended dilution: Flow Cytometry: 1:5-1:10;

Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: Monoclonal
Clone: 3H885
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR