Anti-CD8A Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin) [clone: 5F10/5F13]

Lieferant: United States Biological
USBIC2259-81EA 540 EUR
USBIC2259-81
Anti-CD8A Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin) [clone: 5F10/5F13]
Antikörper
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: Monoclonal
Clone: 5F10/5F13
Conjugation: FITC (Fluorescein)/PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1+IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR