Anti-CCRL2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 12K19]

Lieferant: United States Biological
USBIC2099-81EEA 637 EUR
USBIC2099-81E
Anti-CCRL2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 12K19]
Antikörper
Do not freeze!

Type: Primary
Antigen: CCRL2 (chemokine (C-C motif) receptor-like 2)
Clonality: Monoclonal
Clone: 12K19
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR