Anti-AQP8 Chicken polyclonal antibody unconjugated

Lieferant: United States Biological
USBIA3000-36DEA 951 EUR
USBIA3000-36D
Anti-AQP8 Chicken polyclonal antibody unconjugated
Antikörper
Recommended dilution: ELISA: 0.5-1 ?g/ml; Western Blot: 1-10 ?g/ml

Type: Primary
Antigen: AQP8 (aquaporin 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR