Anti-SERPINI2 Goat Polyclonal Antibody

Lieferant: United States Biological
USBI150657EA 727 EUR
USBI150657
Anti-SERPINI2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:64000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: SERPINI2 (serpin peptidase inhibitor, clade I (pancpin), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR