Anti-Oct-4A Mouse Monoclonal Antibody NL®)557 [clone: 653108]

Lieferant: United States Biological
USBI145558EA 681 EUR
USBI145558
Anti-Oct-4A Mouse Monoclonal Antibody NL®)557 [clone: 653108]
Antikörper
Type: Primary
Antigen: Oct-4A
Clonality: monoclonal
Clone: 653108
Conjugation: NL®557
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR