Anti-ZEB2 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI145376EA 441 EUR
USBI145376
Anti-ZEB2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZEB2 (zinc finger E-box binding homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR