Anti-PRMT1 Mouse Monoclonal Antibody [clone: 13k106]

Lieferant: United States Biological
USBI143233EA 420 EUR
USBI143233
Anti-PRMT1 Mouse Monoclonal Antibody [clone: 13k106]
Antikörper
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: PRMT1 (protein arginine methyltransferase 1)
Clonality: monoclonal
Clone: 13k106
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR