Anti-AKT1/2/3 Mouse Monoclonal Antibody [clone: 13k5]

Lieferant: United States Biological
USBI143132EA 464 EUR
USBI143132
Anti-AKT1/2/3 Mouse Monoclonal Antibody [clone: 13k5]
Antikörper
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: AKT1/2/3 (v-akt murine thymoma viral oncogene homolog 1/2/3)
Clonality: monoclonal
Clone: 13k5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR