Anti-Thymocyte Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI138430EA 714 EUR
USBI138430
Anti-Thymocyte Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Thymocyte
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR