Anti-ZNF230 Mouse Polyclonal Antibody

Lieferant: United States Biological
USBI135641EA 529 EUR
USBI135641
Anti-ZNF230 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF230 (zinc finger protein 230)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR