Anti-UHRF1 Mouse Monoclonal Antibody [clone: 3A11]

Lieferant: United States Biological
USBI135082EA 522 EUR
USBI135082
Anti-UHRF1 Mouse Monoclonal Antibody [clone: 3A11]
Antikörper
Recommended dilution: Immunochemistry: 3?g/ml; Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: UHRF1 (ubiquitin-like with PHD and ring finger domains 1)
Clonality: monoclonal
Clone: 3A11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR