Anti-RTN2 Mouse Polyclonal Antibody

Lieferant: United States Biological
USBI132876EA 522 EUR
USBI132876
Anti-RTN2 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RTN2 (reticulon 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR