Anti-RHOXF1 Mouse Monoclonal Antibody [clone: 4D12]

Lieferant: United States Biological
USBI130766EA 529 EUR
USBI130766
Anti-RHOXF1 Mouse Monoclonal Antibody [clone: 4D12]
Antikörper
Type: Primary
Antigen: RHOXF1 (Rhox homeobox family, member 1)
Clonality: monoclonal
Clone: 4D12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR