Anti-HMGB2 Mouse Monoclonal Antibody [clone: 3C7]

Lieferant: United States Biological
USBI127943EA 529 EUR
USBI127943
Anti-HMGB2 Mouse Monoclonal Antibody [clone: 3C7]
Antikörper
Recommended dilution: Immunochemistry: 1. 5 ?g/ml

Type: Primary
Antigen: HMGB2 (high mobility group box 2)
Clonality: monoclonal
Clone: 3C7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR