Anti-GTF3C3 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI127657EA 565 EUR
USBI127657
Anti-GTF3C3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GTF3C3 (general transcription factor IIIC, polypeptide 3, 102kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR