Anti-GPNMB Mouse Monoclonal Antibody [clone: 1A8]

Lieferant: United States Biological
USBI127490EA 522 EUR
USBI127490
Anti-GPNMB Mouse Monoclonal Antibody [clone: 1A8]
Antikörper
Type: Primary
Antigen: GPNMB (glycoprotein (transmembrane) nmb)
Clonality: monoclonal
Clone: 1A8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR