Anti-F11 Mouse Monoclonal Antibody [clone: 2H8]

Lieferant: US Biological
USBI126534EA 522 EUR
USBI126534
Anti-F11 Mouse Monoclonal Antibody [clone: 2H8]
Antikörper
Type: Primary
Antigen: F11 (coagulation factor XI)
Clonality: monoclonal
Clone: 2H8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR