Anti-DDB1 Mouse Monoclonal Antibody [clone: 4C5]

Lieferant: US Biological
USBI125700EA 529 EUR
USBI125700
Anti-DDB1 Mouse Monoclonal Antibody [clone: 4C5]
Antikörper
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: DDB1 (damage-specific DNA binding protein 1, 127kDa)
Clonality: monoclonal
Clone: 4C5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR