Anti-CPSF3 Mouse Monoclonal Antibody [clone: 6E6]

Lieferant: United States Biological
USBI125292EA 522 EUR
USBI125292
Anti-CPSF3 Mouse Monoclonal Antibody [clone: 6E6]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: CPSF3 (cleavage and polyadenylation specific factor 3, 73kDa)
Clonality: monoclonal
Clone: 6000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR