Anti-COL3 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI125197EA 561 EUR
USBI125197
Anti-COL3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: COL3 (collagen, type III)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR