Anti-B3GAT3 Mouse Polyclonal Antibody

Lieferant: United States Biological
USBI123772EA 568 EUR
USBI123772
Anti-B3GAT3 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: B3GAT3 (beta-1,3-glucuronyltransferase 3 (glucuronosyltransferase I))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR