Anti-ALPK3 Mouse Monoclonal Antibody [clone: 4G4]

Lieferant: United States Biological
USBI123254EA 529 EUR
USBI123254
Anti-ALPK3 Mouse Monoclonal Antibody [clone: 4G4]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 1 ?g/ml

Type: Primary
Antigen: ALPK3 (alpha-kinase 3)
Clonality: monoclonal
Clone: 4G4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR