Anti-SIRT1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI045288EA 443 EUR
USBI045288
Anti-SIRT1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SIRT1 (sirtuin 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR