Anti-ANGPTL7 Mouse Monoclonal Antibody [clone: Kairos 397-7]

Lieferant: US Biological
USBI044552EA 473 EUR
USBI044552
Anti-ANGPTL7 Mouse Monoclonal Antibody [clone: Kairos 397-7]
Antikörper
Type: Primary
Antigen: ANGPTL7 (angiopoietin-like 7)
Clonality: Monoclonal
Clone: Kairos 397-7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR