Anti-XCR1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBI043915-BIOTINEA 791 EUR
USBI043915-BIOTIN
Anti-XCR1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: XCR1 (chemokine (C motif) receptor 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR