Anti-TSPAN1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: United States Biological
USBI043372-FITCEA 791 EUR
USBI043372-FITC
Anti-TSPAN1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: TSPAN1 (tetraspanin 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR