Anti-TNFRSF1A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: United States Biological
USBI043076-HRPEA 850 EUR
USBI043076-HRP
Anti-TNFRSF1A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50; Flow Cytometry: 1:10-50

Type: Primary
Antigen: TNFRSF1A (tumor necrosis factor receptor superfamily, member 1A)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR