Anti-ST7 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: United States Biological
USBI042357-HRPEA 791 EUR
USBI042357-HRP
Anti-ST7 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50

Type: Primary
Antigen: ST7 (suppression of tumorigenicity 7)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR