Anti-SAMM50 Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBI041393-BIOTINEA 791 EUR
USBI041393-BIOTIN
Anti-SAMM50 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: SAMM50 (SAMM50 sorting and assembly machinery component)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR