Anti-RGS13 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI041004EA 627 EUR
USBI041004
Anti-RGS13 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: RGS13 (regulator of G-protein signaling 13)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR