Anti-ZFP42 Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBI040971-BIOTINEA 791 EUR
USBI040971-BIOTIN
Anti-ZFP42 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: ZFP42 (ZFP42 zinc finger protein)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR