Anti-RAMP3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: United States Biological
USBI040819-HRPEA 850 EUR
USBI040819-HRP
Anti-RAMP3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: RAMP3 (receptor (G protein-coupled) activity modifying protein 3)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse