Anti-PIK3R2 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI040052EA 587 EUR
USBI040052
Anti-PIK3R2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: PIK3R2 (phosphoinositide-3-kinase, regulatory subunit 2 (beta))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR