Anti-MMD2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
USBI038494-APCEA 852 EUR
USBI038494-APC
Anti-MMD2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MMD2 (monocyte to macrophage differentiation-associated 2)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse