Anti-MAP1LC3C Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: US Biological
038116-FITC
USBI038116-FITCEA 852 EUR
USBI038116-FITC
Anti-MAP1LC3C Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-100
Do not freeze!

Type: Primary
Antigen: MAP1LC3C (microtubule-associated protein 1 light chain 3 gamma)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR