Anti-LTK Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: United States Biological
USBI038005-APCEA 791 EUR
USBI038005-APC
Anti-LTK Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: LTK (leukocyte receptor tyrosine kinase)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR