Anti-LSM14A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: United States Biological
USBI037995-FITCEA 852 EUR
USBI037995-FITC
Anti-LSM14A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: LSM14A (LSM14A, SCD6 homolog A (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR