Anti-PPP1R37 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: US Biological
037971-AP
USBI037971-APEA 852 EUR
USBI037971-AP
Anti-PPP1R37 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: PPP1R37 (protein phosphatase 1, regulatory subunit 37)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR