Anti-LILRB1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: US Biological
037777-AP
USBI037777-APEA 852 EUR
USBI037777-AP
Anti-LILRB1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: LILRB1 (leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 1)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR