Anti-LHFPL1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: US Biological
037764-APC
USBI037764-APCEA 852 EUR
USBI037764-APC
Anti-LHFPL1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: LHFPL1 (lipoma HMGIC fusion partner-like 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR