Anti-LEMD2 Rabbit Polyclonal Antibody

Lieferant: US Biological
037733
USBI037733EA 587 EUR
USBI037733
Anti-LEMD2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: LEMD2 (LEM domain containing 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR