Anti-KRT79 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: US Biological
USBI037632-APCEA 791 EUR
USBI037632-APC
Anti-KRT79 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: KRT79 (keratin 79)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR