Anti-KLKB1 Rabbit Polyclonal Antibody

Lieferant: US Biological
037574
USBI037574EA 587 EUR
USBI037574
Anti-KLKB1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: KLKB1 (kallikrein B, plasma (Fletcher factor) 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR