Anti-KLF4 Rabbit Polyclonal Antibody

Lieferant: US Biological
037533
USBI037533EA 587 EUR
USBI037533
Anti-KLF4 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Dot Blot 1:50-100

Type: Primary
Antigen: KLF4 (Kruppel-like factor 4 (gut))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR