Anti-KIT Rabbit Polyclonal Antibody

Lieferant: US Biological
037505
USBI037505EA 587 EUR
USBI037505
Anti-KIT Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Dot blot 1:500

Type: Primary
Antigen: KIT (v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR