Anti-KCTD8 Rabbit Polyclonal Antibody

Lieferant: US Biological
037366
USBI037366EA 587 EUR
USBI037366
Anti-KCTD8 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: KCTD8 (potassium channel tetramerization domain containing 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR