Anti-HSPA5 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: US Biological
USBI036803-APEA 791 EUR
USBI036803-AP
Anti-HSPA5 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50
Do not freeze!

Type: Primary
Antigen: HSPA5 (heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa))
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR