Anti-HIST1H2AG Rabbit Polyclonal Antibody

Lieferant: US Biological
USBI036588EA 587 EUR
USBI036588
Anti-HIST1H2AG Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: HIST1H2AG (histone cluster 1, H2ag)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR